Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003918.hg.1 |
ENST00000284984.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003918.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003918.hg.1 |
TC21000345.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003914.hg.1 |
ENST00000284984.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003914.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003914.hg.1 |
TC21000345.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000451462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000517452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000517777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
TC2100007821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000451462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000517452.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
ENST00000517777.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152725.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152731.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152731.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152731.hg.1 |
TC2100007821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100152731.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152731.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152729.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152729.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152729.hg.1 |
TC2100007821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100152729.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152729.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152730.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152730.hg.1 |
NM_006988.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152730.hg.1 |
TC2100007821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100152730.hg.1 |
ENST00000284984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152730.hg.1 |
NM_006988.4 |
Refseq |