Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016302.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016302.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016301.hg.1 |
ENST00000533594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016301.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016301.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016297.hg.1 |
ENST00000533594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016297.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016297.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016300.hg.1 |
ENST00000455942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016300.hg.1 |
ENST00000533594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016300.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016300.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016298.hg.1 |
ENST00000455942.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016298.hg.1 |
ENST00000533594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016298.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016298.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016303.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11016303.hg.1 |
TC11001285.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100174146.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174146.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174146.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174146.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174146.hg.1 |
TC1100009814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000526203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009814.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174145.hg.1 |
TC1100009814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174147.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174147.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174147.hg.1 |
TC1100009814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174143.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174143.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174143.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174143.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174143.hg.1 |
TC1100009814.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
ENST00000455942.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
ENST00000625099.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
NM_007105.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100174144.hg.1 |
TC1100009814.hg.1 |
Clariom D TC |