Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014253.hg.1 |
ENST00000324873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014253.hg.1 |
ENST00000395641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014253.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014253.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014253.hg.1 |
TC16000989.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009865.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000324873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000395641.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
ENST00000567646.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
NM_001042483.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
NM_012385.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175941.hg.1 |
TC1600009865.hg.1 |
Clariom D TC |