Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000292147.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000594342.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000595115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000595115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000595115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000595115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000598330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000600651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000602138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000602138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000602138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000602138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
ENST00000602138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010851.hg.1 |
NM_014297.4 |
Refseq |