Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014369.3 | NP_055184.2 | 5 - 6 | 521 | 64 | |
XM_006712417.1 | XP_006712480.1 | 5 - 6 | 516 | 64 | ||
GenBank mRNA | X79568.1 | - | 5 - 6 | 463 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Homo sapiens 1700 Inventoried Probe spans exons |