Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026571.hg.1 |
TC05001922.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026571.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026571.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026569.hg.1 |
TC05001922.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026569.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026569.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026569.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026570.hg.1 |
TC05001922.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026570.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026570.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186515.hg.1 |
TC0500012420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500186518.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186518.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186518.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186518.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186518.hg.1 |
TC0500012420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000261796.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000505432.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
ENST00000518814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012420.hg.1 |
NM_014443.2 |
Refseq |