Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000156476.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000335422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000335422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000335422.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000376799.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000441527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000595793.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000596955.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000601975.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
ENST00000602090.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011236.hg.1 |
NM_015596.2 |
Refseq |