Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000882.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000882.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000882.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000882.hg.1 |
TC13000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000880.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000880.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000880.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000880.hg.1 |
TC13000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000877.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000877.hg.1 |
TC13000091.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010192.hg.1 |
ENST00000422893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000879.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000879.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000879.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13000879.hg.1 |
TC13000091.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145813.hg.1 |
TC1300006729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300006729.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145807.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145807.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145807.hg.1 |
TC1300006729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012675.hg.1 |
ENST00000605793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145809.hg.1 |
TC1300006729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
ENST00000380842.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
ENST00000460403.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
NM_015932.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300145810.hg.1 |
TC1300006729.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010192.hg.1 |
ENST00000422893.2 |
Ensembl |