Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001677.hg.1 |
TC02000146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001677.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001677.hg.1 |
ENST00000462003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001677.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007015.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000264710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000462003.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000473035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
ENST00000495146.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
NM_016131.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147753.hg.1 |
TC0200007015.hg.1 |
Clariom D TC |