Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016217.2 | NP_057301.1 | 2 - 3 | 1601 | 76 | |
GenBank mRNA | AB033492.1 | - | 2 - 3 | 1601 | 76 | |
BC070068.1 | - | 2 - 3 | 1609 | 30340910 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons |