Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014073.hg.1 |
ENST00000216446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014073.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014073.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014073.hg.1 |
TC14001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014072.hg.1 |
ENST00000216446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014072.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014072.hg.1 |
TC14001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014075.hg.1 |
ENST00000216446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014075.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014075.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014075.hg.1 |
TC14001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014071.hg.1 |
ENST00000216446.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014071.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014071.hg.1 |
TC14001238.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169898.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000556532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009481.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169904.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169904.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169904.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169904.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169904.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169901.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000554395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169895.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000553387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
ENST00000555803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169897.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
ENST00000216446.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
ENST00000557388.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
NM_016445.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169903.hg.1 |
TC1400009481.hg.1 |
Clariom D TC |