Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003707.hg.1 |
TC07003322.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003707.hg.1 |
ENST00000199447.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003707.hg.1 |
ENST00000440017.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07003707.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700196707.hg.1 |
TC0700013355.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000199447.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000426106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000440017.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000444718.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000455500.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
ENST00000476435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013355.hg.1 |
NM_016616.4 |
Refseq |