Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016058.hg.1 |
TC08001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016058.hg.1 |
ENST00000262644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016058.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016062.hg.1 |
TC08001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016062.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016060.hg.1 |
TC08001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016060.hg.1 |
ENST00000262644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016060.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016057.hg.1 |
TC08001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016057.hg.1 |
ENST00000262644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016057.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016059.hg.1 |
TC08001238.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016059.hg.1 |
ENST00000262644.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016059.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171548.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171548.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171548.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171542.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171542.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171542.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171542.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171542.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171546.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171546.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171546.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171546.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171546.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171545.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171545.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171545.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171545.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171545.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
ENST00000520392.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010524.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171544.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
ENST00000262644.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
ENST00000517461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
NM_017813.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800171543.hg.1 |
TC0800010524.hg.1 |
Clariom D TC |