Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028239.hg.1 |
TC01001814.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028239.hg.1 |
ENST00000302637.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028239.hg.1 |
ENST00000366922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01028239.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000467924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000488777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011648.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
ENST00000366922.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
ENST00000490891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
NM_018060.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185744.hg.1 |
TC0100011648.hg.1 |
Clariom D TC |