Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010356.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010356.hg.1 |
ENST00000549260.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010356.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010356.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12010356.hg.1 |
TC12000812.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000550711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
ENST00000550711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164164.hg.1 |
TC1200008683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200164163.hg.1 |
TC1200008683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000303694.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000546689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000547956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000549260.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000550711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
ENST00000550711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_001173982.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008683.hg.1 |
NM_018413.5 |
Refseq |