Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001495.hg.1 |
TC6_mcf_hap5000075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001495.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001554.hg.1 |
TC6_ssto_hap7000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001554.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004625.hg.1 |
TC06000399.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004625.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004625.hg.1 |
ENST00000395388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004625.hg.1 |
ENST00000462200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004625.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001320.hg.1 |
TC6_mann_hap4000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001320.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001729.hg.1 |
TC6_cox_hap2000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001729.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001724.hg.1 |
TC6_dbb_hap3000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001724.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001720.hg.1 |
TC6_dbb_hap3000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001720.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001723.hg.1 |
TC6_dbb_hap3000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001723.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001685.hg.1 |
TC6_qbl_hap6000081.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001685.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001689.hg.1 |
TC6_qbl_hap6000081.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001689.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001550.hg.1 |
TC6_ssto_hap7000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001550.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004626.hg.1 |
TC06000399.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004626.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004626.hg.1 |
ENST00000395388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004626.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001725.hg.1 |
TC6_cox_hap2000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001725.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001316.hg.1 |
TC6_mann_hap4000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001316.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001494.hg.1 |
TC6_mcf_hap5000075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001494.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001690.hg.1 |
TC6_qbl_hap6000081.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001690.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001549.hg.1 |
TC6_ssto_hap7000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001549.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004620.hg.1 |
TC06000399.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004620.hg.1 |
ENST00000395388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06004620.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001719.hg.1 |
TC6_dbb_hap3000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001719.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001317.hg.1 |
TC6_mann_hap4000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001317.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001499.hg.1 |
TC6_mcf_hap5000075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001499.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001724.hg.1 |
TC6_cox_hap2000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001724.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001321.hg.1 |
TC6_mann_hap4000074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001321.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001728.hg.1 |
TC6_cox_hap2000090.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001728.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001498.hg.1 |
TC6_mcf_hap5000075.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001498.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001686.hg.1 |
TC6_qbl_hap6000081.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001686.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001553.hg.1 |
TC6_ssto_hap7000076.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001553.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152823.hg.1 |
TC0600007650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152821.hg.1 |
TC0600007650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152825.hg.1 |
TC0600007650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007650.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152820.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152820.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152820.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152820.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152820.hg.1 |
TC0600007650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
ENST00000374982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
ENST00000395388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
NM_019111.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600152824.hg.1 |
TC0600007650.hg.1 |
Clariom D TC |