Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015656.hg.1 |
ENST00000229335.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015656.hg.1 |
ENST00000537228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015656.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015656.hg.1 |
TC12001180.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015657.hg.1 |
ENST00000229335.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015657.hg.1 |
ENST00000537228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015657.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015657.hg.1 |
TC12001180.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009831.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000543081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000544516.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000545512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173916.hg.1 |
TC1200009831.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173921.hg.1 |
TC1200009831.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000229335.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000537228.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
ENST00000545576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
NM_020661.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200173920.hg.1 |
TC1200009831.hg.1 |
Clariom D TC |