Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000480457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000480457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008167.hg.1 |
TC03000376.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008167.hg.1 |
ENST00000295962.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008167.hg.1 |
ENST00000478253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008167.hg.1 |
ENST00000511761.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03008167.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300155003.hg.1 |
TC0300007606.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000295962.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000463756.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000470440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000475982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000478253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000480457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000480457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000485900.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
ENST00000613521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007606.hg.1 |
NM_020676.6 |
Refseq |