Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000547409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000547409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000336854.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000547409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000547409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000548915.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000550604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552544.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000552947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
ENST00000553049.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_001098673.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007654.hg.1 |
NM_021934.4 |
Refseq |