Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012218.hg.1 |
TC03000685.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012218.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012218.hg.1 |
ENST00000480450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012218.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012219.hg.1 |
TC03000685.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012219.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012219.hg.1 |
ENST00000480450.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03012219.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162208.hg.1 |
TC0300008750.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008750.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
ENST00000436022.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
ENST00000481536.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
NM_024768.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300162207.hg.1 |
TC0300008750.hg.1 |
Clariom D TC |