Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012412.hg.1 |
ENST00000380419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012412.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012412.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012412.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012412.hg.1 |
TC10000993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012414.hg.1 |
ENST00000380419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012414.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012414.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012414.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012414.hg.1 |
TC10000993.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012413.hg.1 |
ENST00000380419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012413.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012413.hg.1 |
TC10000993.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166019.hg.1 |
TC1000009665.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009665.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166020.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166020.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166020.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166020.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166020.hg.1 |
TC1000009665.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
ENST00000380419.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
ENST00000479328.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
NM_001171864.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
NM_024803.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166022.hg.1 |
TC1000009665.hg.1 |
Clariom D TC |