Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057036.hg.1 |
TC01003714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057036.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057034.hg.1 |
TC01003714.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057034.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057034.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016977.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228631.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228631.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228631.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228631.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228631.hg.1 |
TC0100016977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228630.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228630.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228630.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228630.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228630.hg.1 |
TC0100016977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100228632.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228632.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228632.hg.1 |
ENST00000241651.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100228632.hg.1 |
NM_002479.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100228632.hg.1 |
TC0100016977.hg.1 |
Clariom D TC |