Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001616.hg.1 |
ENST00000360214.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001616.hg.1 |
ENST00000360938.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001616.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001616.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21001616.hg.1 |
TC21000169.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
TC2100007140.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100148162.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360214.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000360938.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000432278.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
ENST00000456966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_001256295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007140.hg.1 |
NM_005239.5 |
Refseq |