Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000400780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000527328.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528199.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528838.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528842.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000528988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000529630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000529630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000529630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000529630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000530661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000531447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000531447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000531447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000531447.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000532338.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000533240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000534314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
ENST00000614796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009247.hg.1 |
NM_005526.3 |
Refseq |