Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
TC0X000894.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
ENST00000251900.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
ENST00000398048.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
ENST00000442000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011963.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00162977.hg.1 |
TC0X00009185.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000251900.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000398048.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000420857.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
ENST00000491988.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NM_006089.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009185.hg.1 |
NR_033717.1 |
Refseq |