Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_018942_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104137 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8104138 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8104136 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
207353_PM_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11733657_a_at |
- |
- |
Affymetrix Human Genome U219 Array |
11733657_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014074.hg.1 |
TC04001021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014074.hg.1 |
ENST00000400677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014074.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014071.hg.1 |
TC04001021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014071.hg.1 |
ENST00000400677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014071.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014072.hg.1 |
TC04001021.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014072.hg.1 |
ENST00000400677.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04014072.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168654.hg.1 |
TC0400009992.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168653.hg.1 |
TC0400009992.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400168656.hg.1 |
TC0400009992.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000400677.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000506970.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
ENST00000617742.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_001306142.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009992.hg.1 |
NM_018942.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
214386_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
207353_PM_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
207353_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
214386_at |
- |
- |
Human Genome U133A 2.0 Array |
207353_s_at |
- |
- |
Human Genome U133A 2.0 Array |
214386_PM_at |
- |
- |
Human Genome U133A 2.0 Array |
214386_at |
- |
- |
Human Genome U133A 2.0 Array |
207353_PM_s_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_018942_at |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw581971 |
- |
- |
AffymetrixPathwork Diagnostics Pathchip |
pw500557 |
- |
- |
Human Genome U95 Set |
39535_PM_at |
- |
- |
Human Genome U95 Set |
39536_at |
- |
- |
Human Genome U95 Set |
39535_at |
- |
- |
Human Genome U95 Set |
39536_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
214386_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
207353_s_at3 |
- |
- |
Human Genome U133 Set |
207353_s_at |
- |
- |
Human Genome U133 Set |
207353_PM_s_at |
- |
- |
Human Genome U133 Set |
214386_PM_at |
- |
- |
Human Genome U133 Set |
214386_at |
- |
- |