Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016057.hg.1 |
ENST00000261652.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016057.hg.1 |
ENST00000437538.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016057.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17016057.hg.1 |
TC17001181.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700205544.hg.1 |
TC1700012367.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000261652.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000579009.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000581616.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
ENST00000583789.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012367.hg.1 |
NM_012452.2 |
Refseq |