Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
TC03000965.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
ENST00000305586.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
ENST00000357559.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
ENST00000480918.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
NM_001013439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03016706.hg.1 |
NM_005087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000305586.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000357559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000475315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000480918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000482125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
NM_001013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
NM_005087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000305586.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000357559.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000475315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000480918.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
ENST00000482125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
NM_001013439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
NM_005087.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300206030.hg.1 |
TC0300013908.hg.1 |
Clariom D TC |