Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007264.hg.1 |
TC04000499.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007264.hg.1 |
ENST00000306011.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007264.hg.1 |
NM_005172.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008165.hg.1 |
NM_005172.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156170.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156170.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156170.hg.1 |
TC0400008165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400156172.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156172.hg.1 |
NM_005172.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156172.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156172.hg.1 |
NM_005172.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156172.hg.1 |
TC0400008165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
ENST00000306011.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008165.hg.1 |
NM_005172.1 |
Refseq |