Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000590600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
ENST00000590600.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002299.hg.1 |
ENST00000324000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002299.hg.1 |
ENST00000541833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002299.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002299.hg.1 |
TC18000194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002297.hg.1 |
ENST00000324000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002297.hg.1 |
ENST00000541833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002297.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002297.hg.1 |
TC18000194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002298.hg.1 |
ENST00000324000.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002298.hg.1 |
ENST00000541833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002298.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002298.hg.1 |
TC18000194.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
TC1800007394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150707.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150710.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150710.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150710.hg.1 |
TC1800007394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150710.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150710.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000586360.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000590600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
ENST00000590600.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007394.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150706.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150706.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150706.hg.1 |
TC1800007394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150706.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150706.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150708.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150708.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150708.hg.1 |
TC1800007394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150708.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150708.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150709.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150709.hg.1 |
NM_015879.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150709.hg.1 |
TC1800007394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150709.hg.1 |
ENST00000324000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150709.hg.1 |
NM_015879.2 |
Refseq |