Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011195.hg.1 |
ENST00000361871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011195.hg.1 |
ENST00000399753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011195.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011195.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011195.hg.1 |
TC16000766.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011198.hg.1 |
ENST00000361871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011198.hg.1 |
ENST00000399753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011198.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011198.hg.1 |
TC16000766.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010648.hg.1 |
ENST00000396851.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011191.hg.1 |
ENST00000361871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011191.hg.1 |
ENST00000399753.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011191.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011191.hg.1 |
TC16000766.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011200.hg.1 |
ENST00000361871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011200.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011200.hg.1 |
TC16000766.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168822.hg.1 |
TC1600009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168829.hg.1 |
TC1600009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168817.hg.1 |
TC1600009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168826.hg.1 |
TC1600009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1900010648.hg.1 |
ENST00000396851.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168825.hg.1 |
TC1600009074.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000361871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000399753.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000473663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000489198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000564908.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
ENST00000622125.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009074.hg.1 |
NM_016332.3 |
Refseq |