Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284430.1 | NP_001271359.1 | 13 | 3900 | 86 | |
NM_001284431.1 | NP_001271360.1 | 12 | 3784 | 86 | ||
NM_032175.3 | NP_115551.2 | 13 | 3957 | 86 | ||
XM_011543680.2 | XP_011541982.1 | 13 | 4218 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |