Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021963.3 | NP_068798.1 | 1 | 342 | 65 | |
GenBank mRNA | AB027013.1 | - | 1 | 316 | 65 | |
AF136178.1 | - | 1 | 290 | 65 | ||
AK291297.1 | - | 1 | 300 | 65 | ||
AK316569.1 | - | 1 | 342 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon |