Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
ENST00000249079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
ENST00000403026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
ENST00000403305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013027.hg.1 |
TC22000780.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
TC2200008675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200162663.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000249079.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403026.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000403305.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000418863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000422191.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
ENST00000619227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_001207062.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008675.hg.1 |
NM_032561.4 |
Refseq |