Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032068.hg.1 |
TC02004929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032068.hg.1 |
ENST00000233630.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032068.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032068.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032079.hg.1 |
TC02004929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032079.hg.1 |
ENST00000233630.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032079.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032080.hg.1 |
TC02004929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032080.hg.1 |
ENST00000233630.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032080.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195970.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195970.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195970.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195970.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195970.hg.1 |
TC0200013109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000463744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000489914.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013109.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195968.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195968.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195968.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195968.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195968.hg.1 |
TC0200013109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000233630.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000465993.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000475863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000480844.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000496911.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
ENST00000612049.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
NM_032673.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200195961.hg.1 |
TC0200013109.hg.1 |
Clariom D TC |