Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009648.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009648.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009648.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009648.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009642.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009642.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009642.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009642.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009641.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009641.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009641.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009641.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009646.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009646.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009646.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009646.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009640.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009640.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009640.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009640.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009644.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009644.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009644.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009647.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009647.hg.1 |
ENST00000503590.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009647.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009647.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009643.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009643.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009643.hg.1 |
TC20000702.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008650.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160562.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160562.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160562.hg.1 |
TC2000008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160562.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160562.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160560.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160560.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160560.hg.1 |
TC2000008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160560.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160560.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160561.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160561.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160561.hg.1 |
TC2000008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160561.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160561.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160563.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160563.hg.1 |
NM_000361.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160563.hg.1 |
TC2000008650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160563.hg.1 |
ENST00000377103.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160563.hg.1 |
NM_000361.2 |
Refseq |