Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010886.hg.1 |
ENST00000270590.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010886.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19010886.hg.1 |
TC19000766.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008599.hg.1 |
ENST00000601788.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167163.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167163.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167163.hg.1 |
TC1900008600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167163.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167163.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008600.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167162.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167162.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900167162.hg.1 |
TC1900008600.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900167162.hg.1 |
ENST00000270590.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900167162.hg.1 |
NM_001506.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008599.hg.1 |
ENST00000601788.1 |
Ensembl |