Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001228.hg.1 |
TC09000102.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001228.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001228.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001227.hg.1 |
TC09000102.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001227.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09001227.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183565.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183565.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183565.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183565.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183565.hg.1 |
TC0900012113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183562.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183562.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183562.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183562.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183562.hg.1 |
TC0900012113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183564.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183564.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183564.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183564.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183564.hg.1 |
TC0900012113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183563.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183563.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183563.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183563.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183563.hg.1 |
TC0900012113.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
ENST00000380205.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012113.hg.1 |
NM_002170.3 |
Refseq |