Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006157.hg.1 |
ENST00000327809.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006157.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006157.hg.1 |
TC19000467.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006153.hg.1 |
ENST00000327809.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006153.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006153.hg.1 |
TC19000467.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158173.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158173.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158173.hg.1 |
TC1900007852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158173.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158173.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000454971.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007853.hg.1 |
ENST00000597214.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
TC1900007852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158177.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158174.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158174.hg.1 |
TC1900007852.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158174.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000327809.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
ENST00000594310.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007852.hg.1 |
NM_005304.4 |
Refseq |