Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000381151.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000399312.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000477091.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000482679.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000483977.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
ENST00000488492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_006933.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008496.hg.1 |
NM_032476.3 |
Refseq |