Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010239.hg.1 |
ENST00000169565.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010239.hg.1 |
ENST00000307139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010239.hg.1 |
ENST00000533854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010239.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010239.hg.1 |
TC17000788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010238.hg.1 |
ENST00000533854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010238.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010238.hg.1 |
TC17000788.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010240.hg.1 |
ENST00000169565.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010240.hg.1 |
ENST00000307139.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010240.hg.1 |
ENST00000533854.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010240.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17010240.hg.1 |
TC17000788.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008673.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164787.hg.1 |
TC1700008673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164788.hg.1 |
TC1700008673.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
ENST00000307139.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
ENST00000533854.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
NM_145811.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700164789.hg.1 |
TC1700008673.hg.1 |
Clariom D TC |