Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024001.hg.1 |
ENST00000227471.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024001.hg.1 |
ENST00000530331.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024001.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024001.hg.1 |
TC11001999.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100187654.hg.1 |
TC1100011367.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000227471.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000524455.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000525368.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000528423.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000530138.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000531152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000533424.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000610659.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
ENST00000622364.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011367.hg.1 |
NM_030930.3 |
Refseq |