Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7978057 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7978063 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7978062 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7978060 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7978061 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7978058 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7978056 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7978059 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
231946_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
207699_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010717.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010717.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010717.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010717.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010718.hg.1 |
ENST00000419474.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010718.hg.1 |
ENST00000560206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010718.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14010718.hg.1 |
TC14000961.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163527.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163528.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163528.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163528.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163528.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163528.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163526.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163526.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163526.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163526.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163526.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163535.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163529.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163531.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163531.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163531.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163531.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163531.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400163532.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163532.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163532.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400163532.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400163532.hg.1 |
TC1400008722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000412565.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000419474.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000555334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000606808.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
ENST00000615307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008722.hg.1 |
NM_033400.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
231946_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
231946_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_033400_at |
- |
- |
Human Genome U95 Set |
32013_at |
- |
- |
Human Genome U95 Set |
32013_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231946_at9 |
- |
- |
Human Genome U95 Set |
88943_at |
- |
- |
Human Genome U95 Set |
88943_PM_at |
- |
- |
Human Genome U133 Set |
231946_at |
- |
- |
Human Genome U133 Set |
231946_PM_at |
- |
- |