Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026496.hg.1 |
ENST00000440572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026496.hg.1 |
ENST00000524717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026496.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026496.hg.1 |
TC11002211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026497.hg.1 |
ENST00000524717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026497.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026497.hg.1 |
TC11002211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026494.hg.1 |
ENST00000440572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026494.hg.1 |
ENST00000524717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026494.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026494.hg.1 |
TC11002211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026498.hg.1 |
ENST00000440572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026498.hg.1 |
ENST00000524717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026498.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026498.hg.1 |
TC11002211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008854.hg.1 |
ENST00000582890.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008854.hg.1 |
NR_036125.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026495.hg.1 |
ENST00000440572.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026495.hg.1 |
ENST00000524717.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026495.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026495.hg.1 |
TC11002211.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192764.hg.1 |
TC1100012037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192767.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192767.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192767.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192767.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192767.hg.1 |
TC1100012037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192768.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192768.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192768.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192768.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192768.hg.1 |
TC1100012037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100008854.hg.1 |
ENST00000582890.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008854.hg.1 |
NR_036125.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192766.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192766.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192766.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192766.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192766.hg.1 |
TC1100012037.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000524717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
ENST00000618849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012037.hg.1 |
NM_032427.3 |
Refseq |