Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
229246_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
239384_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
226419_PM_s_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000577267.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000582096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012436.hg.1 |
ENST00000585065.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
239384_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
226419_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
229246_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
226419_PM_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
229246_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
239384_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226419_s_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
239384_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
229246_at11 |
- |
- |
Human Genome U95 Set |
43496_at |
- |
- |
Human Genome U95 Set |
50952_at |
- |
- |
Human Genome U95 Set |
43497_g_at |
- |
- |
Human Genome U95 Set |
43497_PM_g_at |
- |
- |
Human Genome U95 Set |
50952_PM_at |
- |
- |
Human Genome U95 Set |
43496_PM_at |
- |
- |
Human Genome U95 Set |
63834_PM_at |
- |
- |
Human Genome U95 Set |
63834_at |
- |
- |
Human Genome U95 Set |
82512_at |
- |
- |
Human Genome U95 Set |
82512_PM_at |
- |
- |
Human Genome U133 Set |
226419_s_at |
- |
- |
Human Genome U133 Set |
229246_at |
- |
- |
Human Genome U133 Set |
239384_PM_at |
- |
- |
Human Genome U133 Set |
239384_at |
- |
- |
Human Genome U133 Set |
226419_PM_s_at |
- |
- |
Human Genome U133 Set |
229246_PM_at |
- |
- |