Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032242.3 | NP_115618.3 | 31 | 5848 | 115 | |
XM_011512908.2 | XP_011511210.1 | 32 | 5950 | 115 | ||
XM_011512909.1 | XP_011511211.1 | 32 | 6543 | 115 | ||
GenBank mRNA | AK055605.1 | - | 1 | 256 | 115 | |
AK126101.1 | - | 9 | 3040 | 115 | ||
AK127254.1 | - | 1 | 1020 | 115 | ||
AK128612.1 | - | 5 | 985 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon |