Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017477.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017477.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017477.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017478.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017478.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017478.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017481.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017481.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017481.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017482.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017482.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017482.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017484.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017484.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017484.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017479.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017479.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017479.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017480.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017480.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14017480.hg.1 |
TC14001562.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176804.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176800.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176802.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176810.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176810.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176810.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176810.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176810.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176803.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176798.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176805.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400176809.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176809.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176809.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400176809.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400176809.hg.1 |
TC1400010390.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000333244.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000555544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
ENST00000557457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010390.hg.1 |
NM_138420.3 |
Refseq |