Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018187.hg.1 |
ENST00000308251.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018187.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018187.hg.1 |
TC16001311.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018189.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018189.hg.1 |
TC16001311.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018188.hg.1 |
ENST00000308251.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018188.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018188.hg.1 |
TC16001311.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185923.hg.1 |
TC1600011050.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185924.hg.1 |
TC1600011050.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600185926.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185926.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185926.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185926.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185926.hg.1 |
TC1600011050.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011050.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
ENST00000308251.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
ENST00000568181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
NM_172347.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600185925.hg.1 |
TC1600011050.hg.1 |
Clariom D TC |