Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000250263.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000518663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000518663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000518663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000518663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000518663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000519292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520332.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000520684.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000521844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000521844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000521844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000521844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000521844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522258.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000522612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
ENST00000523898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006663.hg.1 |
NM_153332.3 |
Refseq |