Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018766.hg.1 |
TC0X001412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018766.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018766.hg.1 |
ENST00000433339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018766.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018770.hg.1 |
TC0X001412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018770.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018770.hg.1 |
ENST00000433339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018770.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018768.hg.1 |
TC0X001412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018768.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018768.hg.1 |
ENST00000433339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018768.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018771.hg.1 |
TC0X001412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018771.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018771.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018769.hg.1 |
TC0X001412.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018769.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018769.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174227.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174227.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174227.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174227.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174227.hg.1 |
TC0X00010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010921.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174223.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174223.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174223.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174223.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174223.hg.1 |
TC0X00010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00174225.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174225.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174225.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174225.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174225.hg.1 |
TC0X00010921.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00174226.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174226.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174226.hg.1 |
ENST00000305963.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174226.hg.1 |
NM_173470.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174226.hg.1 |
TC0X00010921.hg.1 |
Clariom D TC |